Anti-PSAT1 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB25307-1EA 0 EUR
BIRBORB25307-1
Anti-PSAT1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR