Anti-ALP Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB25359-50EA 0 EUR
BIRBORB25359-50
Anti-ALP Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:3000-1:15000

Type: Primary
Antigen: ALP (Alkaline Phosphatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR