Anti-Peroxidase (HRP) Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Biorbyt
BIRBORB25363-50EA 0 EUR
BIRBORB25363-50
Anti-Peroxidase (HRP) Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
ELISA: 1:1000-1:5000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: Peroxidase (HRP)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR