Anti-c-Rel subunit Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB25380-100EA 0 EUR
BIRBORB25380-100
Anti-c-Rel subunit Rabbit Polyclonal Antibody
Antikörper
DB: 1:1000

Type: Primary
Antigen: c-Rel subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR