Anti-PEP Carboxylase Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB25412-2EA 0 EUR
BIRBORB25412-2
Anti-PEP Carboxylase Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:30000-1:160000

Type: Primary
Antigen: PEP Carboxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR