Anti-MPO Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB25566-1EA 0 EUR
BIRBORB25566-1
Anti-MPO Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR