Anti-TF Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Biorbyt
BIRBORB25733-1EA 0 EUR
BIRBORB25733-1
Anti-TF Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
EIA: 1:3200-1:6400

Type: Primary
Antigen: TF (transferrin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR