Anti-Oligodendrocytes + Myelin Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB25851-125EA 0 EUR
BIRBORB25851-125
Anti-Oligodendrocytes + Myelin Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 1:50

Type: Primary
Antigen: Oligodendrocytes + Myelin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR