Anti-COL5 Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB25939-100EA 0 EUR
BIRBORB25939-100
Anti-COL5 Mouse Monoclonal Antibody
Antikörper
  • ELISA: 1-3; DB: 2-4

Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR