Anti-Sm. Cell Lung Carcinoma Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB25992-0.5EA 0 EUR
BIRBORB25992-0.5
Anti-Sm. Cell Lung Carcinoma Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Sm. Cell Lung Carcinoma
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR