Anti-CD58 Mouse Monoclonal Antibody (Biotin)

Lieferant: Biorbyt
BIRBORB26124-100EA 0 EUR
BIRBORB26124-100
Anti-CD58 Mouse Monoclonal Antibody (Biotin)
Antikörper
Flow Cytometry: 1:200

Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR