Anti-SNCA Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB26663-0.2EA 0 EUR
BIRBORB26663-0.2
Anti-SNCA Mouse Monoclonal Antibody
Antikörper
Western Blot: 1:100-1:10000; Immunochemistry: 1:100-1:1000

Type: Primary
Antigen: SNCA (synuclein, alpha (non A4 component of amyloid precursor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR