Anti-APP Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB26679-0.2EA 0 EUR
BIRBORB26679-0.2
Anti-APP Goat Polyclonal Antibody
Antikörper
ELISA: 1:1500; Immunochemistry: 1:100; Immunochemistry: 1:5000; Western Blot: 1:1000

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR