Anti-APP Mouse Monoclonal Antibody (Biotin)

Lieferant: Biorbyt
BIRBORB26757-100EA 0 EUR
BIRBORB26757-100
Anti-APP Mouse Monoclonal Antibody (Biotin)
Antikörper
ELISA: 0.05 ?g/ml; Western Blot: 0.5 ?g/ml; Immunochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR