Anti-AGE-4 Methylglyoxal modifiied Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB27287-10EA 0 EUR
BIRBORB27287-10
Anti-AGE-4 Methylglyoxal modifiied Mouse Monoclonal Antibody
Antikörper
Western Blot: 0.1 ?g/ml; ELISA: 0.05 ?g/ml

Type: Primary
Antigen: AGE-4 Methylglyoxal modifiied
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR