Anti-COL4 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB27319-0.1EA 0 EUR
BIRBORB27319-0.1 BIRBORB27319-0.5
Anti-COL4 Rabbit Polyclonal Antibody
Antikörper
ImmunofluorescenceA 1:40-80; Immunochemistry: 1:500 - 1:1000; ELISA: > 1:200

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR