Anti-COL5 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB27324-0.5EA 0 EUR
BIRBORB27324-0.5
Anti-COL5 Rabbit Polyclonal Antibody
Antikörper
ImmunofluorescenceA 1:40; Immunochemistry: 1:200 - 1:600

Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR