Anti-PCNA Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB27447-5EA 0 EUR
BIRBORB27447-5
Anti-PCNA Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR