Anti-CKAP4 Mouse monoclonal antibody

Lieferant: Biorbyt
BIRBORB27455-5EA 0 EUR
BIRBORB27455-5
Anti-CKAP4 Mouse monoclonal antibody
Antikörper
Type: Primary
Antigen: CKAP4 (cytoskeleton-associated protein 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR