Anti-Fibronectin Rabbit polyclonal antibody

Lieferant: Biorbyt
BIRBORB27486-0.1EA 0 EUR
BIRBORB27486-0.1 BIRBORB27486-0.5
Anti-Fibronectin Rabbit polyclonal antibody
Antikörper
ImmunofluorescenceA 1:80; Immunochemistry: 1:1000 (polymer technique); ELISA: 1:200

Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR