Anti-CSF2 Rabbit polyclonal antibody

Lieferant: Biorbyt
BIRBORB27647-100EA 0 EUR
BIRBORB27647-100
Anti-CSF2 Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: CSF2 (colony stimulating factor 2 (granulocyte-macrophage))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR