Anti-IgG Mouse Antibody

Lieferant: Biorbyt
BIRBORB27709-0.5EA 0 EUR
BIRBORB27709-0.5 BIRBORB27709-1
Anti-IgG Mouse Antibody
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR