Anti-ATRX Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB40826-100EA 0 EUR
BIRBORB40826-100
Anti-ATRX Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: ATRX (alpha thalassemia/mental retardation syndrome X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR