Anti-Dia 1 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB40895-100EA 0 EUR
BIRBORB40895-100
Anti-Dia 1 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: Dia 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human