Anti-Tubby related protein 1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB41024-100EA 0 EUR
BIRBORB41024-100
Anti-Tubby related protein 1 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:500-1:3000

Type: Primary
Antigen: Tubby related protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR