Anti-TRIOBP Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB41574-100EA 0 EUR
BIRBORB41574-100
Anti-TRIOBP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIOBP (TRIO and F-actin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR