Anti-MCM7 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB41616-100EA 0 EUR
BIRBORB41616-100
Anti-MCM7 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: MCM7 (minichromosome maintenance complex component 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR