Anti-PCDH1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB41871-100EA 0 EUR
BIRBORB41871-100
Anti-PCDH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PCDH1 (protocadherin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR