Anti-HDAC3 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB43498-100EA 0 EUR
BIRBORB43498-100
Anti-HDAC3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR