Anti-SDC1 Mouse Monoclonal Antibody (Peridinin Chlorophyll)

Lieferant: Biorbyt
BIRBORB43671-100EA 0 EUR
BIRBORB43671-100
Anti-SDC1 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Antikörper
Type: Primary
Antigen: SDC1 (syndecan 1)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR