Anti-HLA-A2 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Lieferant: Biorbyt
BIRBORB43677-100EA 0 EUR
BIRBORB43677-100
Anti-HLA-A2 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antikörper
Type: Primary
Antigen: HLA-A2
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR