Anti-Cytokeratin Mouse monoclonal antibody
Lieferant: Biorbyt
BIRBORB43709-0.1EA 0 EUR
BIRBORB43709-0.1
Anti-Cytokeratin Mouse monoclonal antibody
Antikörper
Type: Primary
Antigen: Cytokeratin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen