Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein))

Lieferant: Biorbyt
BIRBORB43739-100EA 0 EUR
BIRBORB43739-100
Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR