Anti-HLA-DR Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB43864-0.1EA 0 EUR
BIRBORB43864-0.1
Anti-HLA-DR Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR