Anti-CD98 Mouse Monoclonal Antibody (Biotin)

Lieferant: Biorbyt
BIRBORB44026-0.1EA 0 EUR
BIRBORB44026-0.1
Anti-CD98 Mouse Monoclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CD98
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR