Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Lieferant: Biorbyt
BIRBORB44373-100EA 0 EUR
BIRBORB44373-100
Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antikörper
Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR