Anti-PRR7 Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB44753-0.1EA 0 EUR
BIRBORB44753-0.1
Anti-PRR7 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PRR7 (proline rich 7 (synaptic))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR