Anti-CD4 Rat Monoclonal Antibody (FITC (Fluorescein))

Lieferant: Biorbyt
BIRBORB44809-0.1EA 0 EUR
BIRBORB44809-0.1
Anti-CD4 Rat Monoclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR