Anti-ITGAL Rat Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB44825-0.1EA 0 EUR
BIRBORB44825-0.1
Anti-ITGAL Rat Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR