Anti-EGFR E746-A750 deletion Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB47909-100EA 0 EUR
BIRBORB47909-100
Anti-EGFR E746-A750 deletion Mouse Monoclonal Antibody
Antikörper
Western Blot: 1:500-1000

Type: Primary
Antigen: EGFR E746-A750 deletion
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR