Anti-AR Rabbit polyclonal antibody

Lieferant: Biorbyt
BIRBORB47963-100EA 0 EUR
BIRBORB47963-100
Anti-AR Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: AR (androgen receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR