Anti-MAP3K2 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB47966-100EA 0 EUR
BIRBORB47966-100
Anti-MAP3K2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP3K2 (mitogen-activated protein kinase kinase kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR