Anti-CASP8 Rabbit polyclonal antibody
Lieferant: Biorbyt
BIRBORB47991-100EA 0 EUR
BIRBORB47991-100
Anti-CASP8 Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: CASP8 (caspase 8, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human