Anti-BSG Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB47993-100EA 0 EUR
BIRBORB47993-100
Anti-BSG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR