Anti-HNRNPA2B1 Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB48089-100EA 0 EUR
BIRBORB48089-100
Anti-HNRNPA2B1 Mouse Monoclonal Antibody
Antikörper
Western Blot: 1:1000-1:20000;

Type: Primary
Antigen: HNRNPA2B1 (heterogeneous nuclear ribonucleoprotein A2/B1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR