Anti-AGER Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB48272-100EA 0 EUR
BIRBORB48272-100
Anti-AGER Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AGER (advanced glycosylation end product-specific receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR