Anti-TIGD3 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB126428-200EA 0 EUR
BIRBORB126428-200 BIRBORB126428-100
Anti-TIGD3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIGD3 (tigger transposable element derived 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human