Anti-TR10A Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB126435-100EA 0 EUR
BIRBORB126435-100 BIRBORB126435-200
Anti-TR10A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TR10A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human