Anti-TOP2A Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB126445-200EA 0 EUR
BIRBORB126445-200 BIRBORB126445-100
Anti-TOP2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR