Anti-TP53I11 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB126450-200EA 0 EUR
BIRBORB126450-200 BIRBORB126450-100
Anti-TP53I11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TP53I11 (tumor protein p53 inducible protein 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR